CAS RN 127503-04-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Lithium tantalum ethoxide, 99.9% (metals basis), 5% w/v in ethanol, Thermo Scientific™
CAS: 127503-04-2 MDL Number: MFCD00210630 Synonym: lithium tantalum ethoxide