CAS RN 127913-44-4
CAS RN 127913-44-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(3S)-4-Chloro-3-hydroxybutyronitrile, TRC
CAS: 127913-44-4 Molecular Formula: C4H6ClNO Molecular Weight (g/mol): 119.55 Synonym: (3S)-4-Chloro-3-hydroxybutanenitrile,(S)-4-Chloro-3-hydroxybutanenitrile,(S)-(-)-4-Chloro-3-hydroxybutyronitrile,(S)-4-Chloro-3-hydroxybutyronitrile IUPAC Name: (3S)-4-chloro-3-hydroxybutanenitrile SMILES: O[C@H](CCl)CC#N