accessibility menu, dialog, popup

UserName

CH3NNOOClFFF

CAS RN 128455-63-0

IUPAC Name: 5-chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylate
Synonyms: 5-chloro-1-methyl-3-(trifluoromethyl)pyrazole-4-carboxylate
Molecular Weight (g/mol): 227.55
Molecular Formula: C6H3ClF3N2O2
InChi Key: IKGVBNQPAJOSFP-UHFFFAOYSA-M
SMILES: CN1N=C(C(C([O-])=O)=C1Cl)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results