CAS RN 130047-14-2
CAS RN 130047-14-2
IUPAC Name:
methyl 4-[(chlorosulfonyl)methyl]benzoate
Synonyms:
methyl 4-[(chlorosulfonyl)methyl]benzoate
Molecular Weight (g/mol):
248.68
Molecular Formula:
C9H9ClO4S
InChi Key:
VTQZPLOSFBIFGF-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC=C(CS(Cl)(=O)=O)C=C1
1
–
1
of
1
results
1
Methyl 4-[(chlorosulfonyl)methyl]benzoate, 95%, Thermo Scientific™
CAS: 130047-14-2 Molecular Formula: C9H9ClO4S Molecular Weight (g/mol): 248.68 MDL Number: MFCD07778383 InChI Key: VTQZPLOSFBIFGF-UHFFFAOYSA-N Synonym: methyl 4-chlorosulfonyl methyl benzoate,4-methoxycarbonyl phenyl methanesulphonyl chloride,benzoic acid, 4-chlorosulfonyl methyl-, methyl ester,benzoic acid,4-chlorosulfonyl methyl-, methyl ester,acmc-20dqi1,methyl 4-chlorosulfonylmethyl benzoate,methyl 4-chlorosulphonyl methyl benzoate,4-chlorosulfonylmethylbenzoic acid methyl ester,4-methoxycarbonyl phenyl methanesulfonyl chloride PubChem CID: 21804301 IUPAC Name: methyl 4-(chlorosulfonylmethyl)benzoate SMILES: COC(=O)C1=CC=C(CS(Cl)(=O)=O)C=C1