CAS RN 13005-09-9
CAS RN 13005-09-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dehydrocorydaline nitrate, TRC
CAS: 13005-09-9 Molecular Formula: C22H24N2O7 Molecular Weight (g/mol): 428.4 Synonym: 6H-Dibenzo[a,g]quinolizinium, 5-hydro-2,3,9,10-tetramethoxy-13-methyl-, nitrate (1:1) (ACI),Dibenzo[a,g]quinolizinium, 5,6-dihydro-2,3,9,10-tetramethoxy-13-methyl-, nitrate (9CI),Dehydrocorydaline nitrate,5-Hydro-2,3,9,10-tetramethoxy-13-methyl-6H-dibenzo[a,g]quinolizinium nitrate SMILES: COc1cc2CC[n+]3cc4c(OC)c(OC)ccc4c(C)c3c2cc1OC.[O-][N+](=O)[O-]