accessibility menu, dialog, popup

UserName

CH3NNCH3NH3C

CAS RN 13036-81-2

IUPAC Name: 1,3,5-tripropyl-1,3,5-triazinane
Synonyms: 1,3,5-tripropyl-1,3,5-triazinane
Molecular Weight (g/mol): 213.37
Molecular Formula: C12H27N3
InChi Key: DPHBVWJGMBYPMK-UHFFFAOYSA-N
SMILES: CCCN1CN(CCC)CN(CCC)C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results