CAS RN 13071-11-9
IUPAC Name:
hydrogen (2R)-1-(naphthalen-1-yloxy)-3-[(propan-2-yl)amino]propan-2-ol chloride
Synonyms:
D-propranolol hydrochloride dexpropranolol hcl dexpropranolol hydrochloride
Molecular Weight (g/mol):
295.81
Molecular Formula:
C16H22ClNO2
InChi Key:
ZMRUPTIKESYGQW-PFEQFJNWSA-N
SMILES:
[H+].[Cl-].CC(C)NC[C@@H](O)COC1=C2C=CC=CC2=CC=C1
1
–
2
of
2
results
1
(R)-Propranolol Hydrochloride, TRC
CAS: 13071-11-9 Molecular Formula: C16H22ClNO2 Molecular Weight (g/mol): 295.8 Synonym: (2R)-1-[(1-Methylethyl)amino]-3-(1-naphthalenyloxy)-2-propanol Hydrochloride,(+)-1-(Isopropylamino)-3-(1-naphthyloxy)-2-propanol Hydrochloride,(+)-Anapriline,(+)-Propranolol Hydrochloride,(R)-(+)-Propranolol Hydrochloride,AY 20694,Dexpropranolol Hydrochloride,ICI 47319,d-Anapriline,d-Propranolol Hydrochloride; IUPAC Name: (2R)-1-naphthalen-1-yloxy-3-(propan-2-ylamino)propan-2-ol;hydrochloride SMILES: Cl.C=CCN1C(CCC1)CNC(=O)c2c(OC)cc3c(nnn3)c2