accessibility menu, dialog, popup

UserName

OOODyDy

CAS RN 1308-87-8

IUPAC Name: didysprosium(3+) trioxidandiide
Synonyms: dysprosium(III) oxide
Molecular Weight (g/mol): 373
Molecular Formula: Dy2O3
InChi Key: GEZAXHSNIQTPMM-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[Dy+3].[Dy+3]
Physical Form 
  • (6)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (1)
  • (6)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)