accessibility menu, dialog, popup

UserName

CAS RN 130870-00-7

IUPAC Name: (4-bromo-2,5-dimethylphenyl)boronic acid
Synonyms: 4-bromo-2,5-dimethylphenylboronic acid
Molecular Weight (g/mol): 228.88
Molecular Formula: C8H10BBrO2
InChi Key: FUVZURQKCDUKIR-UHFFFAOYSA-N
SMILES: CC1=CC(B(O)O)=C(C)C=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results