accessibility menu, dialog, popup

UserName

BFFFFH3CH3CCH3PHCH3CH3H3CCH3CH3H3C

CAS RN 131274-22-1

IUPAC Name: tetrafluoroboranuide; tri-tert-butylphosphanium
Synonyms: tri-tert-butylphosphanium tetrafluoroborate
Molecular Weight (g/mol): 290.13
Molecular Formula: C12H28BF4P
InChi Key: YTJUCJAUJCXFTN-UHFFFAOYSA-O
SMILES: F[B-](F)(F)F.CC(C)(C)[PH+](C(C)(C)C)C(C)(C)C
Quantity 
  • (3)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)