CAS RN 1313730-19-6
CAS RN 1313730-19-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
MedChemExpress TFLLR-NH2(TFA)
MedChemExpress TFLLR-NH2 (TFA) is a selective PAR1 agonist with an EC50 of 1.9 μM.
2
MedChemExpress TFLLR-NH2(TFA)
MedChemExpress TFLLR-NH2 (TFA) is a selective PAR1 agonist with an EC50 of 1.9 μM.