accessibility menu, dialog, popup

UserName

CAS RN 13138-74-4

CH3OONHNOO

CAS RN 13138-74-4

IUPAC Name: methyl 4-nitro-1H-pyrrole-2-carboxylate
Synonyms: methyl 4-nitro-1H-pyrrole-2-carboxylate
Molecular Weight (g/mol): 170.12
Molecular Formula: C6H6N2O4
InChi Key: BRZNAATZQZPGBQ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(=CN1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results