accessibility menu, dialog, popup

UserName

OMgBrH3C

CAS RN 13139-86-1

IUPAC Name: magnesium(2+) anisole bromide
Synonyms: magnesium(2+) anisole bromide
Molecular Weight (g/mol): 211.34
Molecular Formula: C7H7BrMgO
InChi Key: RBWRWAUAVRMBAC-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].COC1=CC=[C-]C=C1
Quantity 
  • (3)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results