CAS RN 1314-22-3
IUPAC Name:
zinc peroxide
Synonyms:
zinc peroxide
Molecular Weight (g/mol):
99.39
Molecular Formula:
H2O2Zn
InChi Key:
DLINORNFHVEIFE-UHFFFAOYSA-N
SMILES:
[Zn].OO
1
–
1
of
1
results
1
Zinc peroxide, approximately 50% ZnO{2}
CAS: 1314-22-3 Molecular Formula: O2Zn Molecular Weight (g/mol): 97.38 MDL Number: MFCD00137469 InChI Key: IPTOGCUGCFHDSS-UHFFFAOYSA-N Synonym: zinc peroxide,zinc superoxide,zinc dioxide,zinc peroxide un1516 oxidizer,zinc peroxide, medicinal,unii-0i969dvm77,zinc peroxide, medicinal usp,zpo PubChem CID: 14809 SMILES: [Zn++].[O-][O-]