CAS RN 1314419-65-2
CAS RN 1314419-65-2
IUPAC Name:
1-tert-butyl 3-ethyl pyrrolidine-1,3-dicarboxylate
Synonyms:
1-tert-butyl 3-ethyl pyrrolidine-1,3-dicarboxylate
Molecular Weight (g/mol):
243.3
Molecular Formula:
C12H21NO4
InChi Key:
PKDQVUYUWUHNMG-UHFFFAOYNA-N
SMILES:
CCOC(=O)C1CCN(C1)C(=O)OC(C)(C)C
1
–
1
of
1
results
1
N-Boc-D-beta-proline ethyl ester, 97%, Thermo Scientific™
CAS: 1314419-65-2 Molecular Formula: C12H21NO4 Molecular Weight (g/mol): 243.303 MDL Number: MFCD19442798 InChI Key: PKDQVUYUWUHNMG-SECBINFHSA-N Synonym: ethyl r-1-boc-3-pyrrolidinecarboxylate,r-1-tert-butyl 3-ethyl pyrrolidine-1,3-dicarboxylate,n-boc-d-beta-proline ethyl ester,ethyl r-1-boc-pyrrolidine-3-carboxylate,r-1-boc-pyrrolidine-3-carboxylic acid ethyl ester,r-1-n-boc-pyrrolidine-3-carboxylic acid ethyl ester,1-o-tert-butyl 3-o-ethyl 3r-pyrrolidine-1,3-dicarboxylate,1-tert-butyl 3-ethyl 3r-pyrrolidine-1,3-dicarboxylate PubChem CID: 1515160 IUPAC Name: 1-O-tert-butyl 3-O-ethyl (3R)-pyrrolidine-1,3-dicarboxylate SMILES: CCOC(=O)C1CCN(C1)C(=O)OC(C)(C)C