accessibility menu, dialog, popup

UserName

CAS RN 131534-65-1

OBON

CAS RN 131534-65-1

IUPAC Name: 3-(1,3,2-dioxaborinan-2-yl)pyridine
Synonyms: 3-(1,3,2-dioxaborinan-2-yl)pyridine
Molecular Weight (g/mol): 162.98
Molecular Formula: C8H10BNO2
InChi Key: ZUUZNOFTKGDLLN-UHFFFAOYSA-N
SMILES: C1COB(OC1)C1=CC=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results