CAS RN 13189-89-4
CAS RN 13189-89-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2,3-Dihydroxybenzamide, TRC
CAS: 13189-89-4 Molecular Formula: C7H7NO3 Molecular Weight (g/mol): 153.14 Synonym: 3-Carbamoyl-1,2-benzenediol SMILES: NC(=O)c1cccc(O)c1O