accessibility menu, dialog, popup

UserName

CAS RN 132706-12-8

NSO

CAS RN 132706-12-8

IUPAC Name: 5-(pyridin-2-yl)thiophene-2-carbaldehyde
Synonyms: 5-(pyridin-2-yl)thiophene-2-carbaldehyde
Molecular Weight (g/mol): 189.23
Molecular Formula: C10H7NOS
InChi Key: VAGVVFZWHTULBM-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(S1)C1=CC=CC=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results