CAS RN 13276-17-0
CAS RN 13276-17-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-(Morpholinoethyl)acrylamide, TRC
CAS: 13276-17-0 Molecular Formula: C9H16N2O2 Molecular Weight (g/mol): 184.24 Synonym: N-[2-(4-Morpholinyl)ethyl]-2-propenamide,N-(2-Morpholinoethyl)acrylamide,N-[2-(4-Morpholinyl)ethyl]acrylamide IUPAC Name: N-(2-morpholin-4-ylethyl)prop-2-enamide SMILES: C=CC(=O)NCCN1CCOCC1