CAS RN 1330173-19-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
rac N-Demethyl Promethazine-d3 Hydrochloride, TRC
CAS: 1330173-19-7 Molecular Formula: C16H16D3ClN2S Molecular Weight (g/mol): 309.87 Synonym: N,α-Dimethyl-10H-phenothiazine-10-ethanamine-d3 Hydrochloride,Nor1-Promethazine-d3 Monohydrochloride SMILES: CC(NC([2H])([2H])[2H])CN1C2=C(C=CC=C2)SC3=CC=CC=C31.Cl