accessibility menu, dialog, popup

UserName

ClHGeH2HClCH3H3C

CAS RN 13314-52-8

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 205.69
Molecular Formula: C4H14Cl2Ge
InChi Key: FVUWAWRMMUJVGO-UHFFFAOYSA-N
SMILES: Cl.Cl.CC[GeH2]CC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results