CAS RN 13324-20-4
CAS RN 13324-20-4
IUPAC Name:
1-amino-4-({3-[(4,6-dichloro-1,3,5-triazin-2-yl)amino]-4-sulfophenyl}amino)-9,10-dioxo-9,10-dihydroanthracene-2-sulfonic acid
Synonyms:
procion blue MX-R
Molecular Weight (g/mol):
637.42
Molecular Formula:
C23H14Cl2N6O8S2
InChi Key:
RTLULCVBFCRQKI-UHFFFAOYSA-N
SMILES:
NC1=C(C=C(NC2=CC=C(C(NC3=NC(Cl)=NC(Cl)=N3)=C2)S(O)(=O)=O)C2=C1C(=O)C1=CC=CC=C1C2=O)S(O)(=O)=O
1
–
1
of
1
results
1
Thermo Scientific Chemicals Reactive Blue 4, dye content min. 35%
CAS: 13324-20-4 Molecular Formula: C23H14Cl2N6O8S2 Molecular Weight (g/mol): 637.42 MDL Number: MFCD00001218 InChI Key: RTLULCVBFCRQKI-UHFFFAOYSA-N Synonym: C.I. 61205; Procion™ Blue MX-R PubChem CID: 25863 IUPAC Name: 1-amino-4-[3-[(4,6-dichloro-1,3,5-triazin-2-yl)amino]-4-sulfoanilino]-9,10-dioxoanthracene-2-sulfonic acid SMILES: NC1=C(C=C(NC2=CC=C(C(NC3=NC(Cl)=NC(Cl)=N3)=C2)S(O)(=O)=O)C2=C1C(=O)C1=CC=CC=C1C2=O)S(O)(=O)=O