accessibility menu, dialog, popup

UserName

H3COHN(R)(R)OH(S)O(S)O(R)O(S)O

CAS RN 13343-63-0

IUPAC Name: N-[(2S,4aR,6S,7R,8R,8aS)-6-(benzyloxy)-8-hydroxy-2-phenyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-7-yl]acetamide
Synonyms: N-[(2S,4aR,6S,7R,8R,8aS)-6-(benzyloxy)-8-hydroxy-2-phenyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-7-yl]acetamide
Molecular Weight (g/mol): 399.44
Molecular Formula: C22H25NO6
InChi Key: NXGXFAKJUWEFEC-VXAPCQRLSA-N
SMILES: CC(=O)N[C@@H]1[C@@H](O)[C@@H]2O[C@H](OC[C@H]2O[C@@H]1OCC1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results