accessibility menu, dialog, popup

UserName

CH3OOCH3NH3C(E/Z)

CAS RN 13412-12-9

IUPAC Name: methyl 3-(methylimino)butanoate
Synonyms: methyl 3-(methylimino)butanoate
Molecular Weight (g/mol): 129.16
Molecular Formula: C6H11NO2
InChi Key: UPQBRLACXPZFPJ-UHFFFAOYSA-N
SMILES: COC(=O)CC(C)=NC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results