accessibility menu, dialog, popup

UserName

OSiO

CAS RN 1343-98-2

IUPAC Name: dihydroxysilanone
Synonyms: dihydroxysilanone
Molecular Weight (g/mol): 78.1
Molecular Formula: H2O3Si
InChi Key: IJKVHSBPTUYDLN-UHFFFAOYSA-N
SMILES: O[Si](O)=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)