accessibility menu, dialog, popup

UserName

CH3CH3H3CNHSNHH2N

CAS RN 13431-39-5

IUPAC Name: 3-amino-1-tert-butylthiourea
Synonyms: 3-amino-1-tert-butylthiourea
Molecular Weight (g/mol): 147.24
Molecular Formula: C5H13N3S
InChi Key: ZUWRCNZOBNETMU-UHFFFAOYSA-N
SMILES: CC(C)(C)NC(=S)NN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results