accessibility menu, dialog, popup

UserName

BrBrPd

CAS RN 13444-94-5

IUPAC Name: palladium(2+) dibromide
Synonyms: palladium(II) bromide
Molecular Weight (g/mol): 266.23
Molecular Formula: Br2Pd
InChi Key: INIOZDBICVTGEO-UHFFFAOYSA-L
SMILES: [Br-].[Br-].[Pd++]
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (4)
  • (6)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)