accessibility menu, dialog, popup

UserName

NH4NH4OTeOOO

CAS RN 13453-06-0

IUPAC Name: diammonium tellurate
Synonyms: diammonium tellurate
Molecular Weight (g/mol): 227.67
Molecular Formula: H8N2O4Te
InChi Key: DRGYXGZFRXFMHF-UHFFFAOYSA-N
SMILES: [NH4+].[NH4+].[O-][Te]([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results