accessibility menu, dialog, popup

UserName

CAS RN 13462-93-6

NH4OHAsOHOO

CAS RN 13462-93-6

IUPAC Name: ammonium dihydrogen arsorate
Synonyms: ammonium dihydrogen arsenate
Molecular Weight (g/mol): 158.97
Molecular Formula: AsH6NO4
InChi Key: VTWJXWNBCACAAK-UHFFFAOYSA-N
SMILES: [NH4+].O[As](O)([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results