accessibility menu, dialog, popup

UserName

H2OH2OH2OH2OBrBrCd

CAS RN 13464-92-1

IUPAC Name: cadmium(2+) tetrahydrate dibromide
Synonyms: cadmium(2+) tetrahydrate dibromide
Molecular Weight (g/mol): 344.28
Molecular Formula: Br2CdH8O4
InChi Key: PANYFCXNZSEXSU-UHFFFAOYSA-L
SMILES: O.O.O.O.[Br-].[Br-].[Cd++]
Grade 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)