CAS RN 1346598-70-6
CAS RN 1346598-70-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Desbutyl Dronedarone-d6 Hydrochloride, TRC
CAS: 1346598-70-6 Molecular Formula: C27H30D6N2O5S • HCl Molecular Weight (g/mol): 543.15 Synonym: N-(2-Butyl-3-(4-(3-(butylamino)propoxy-1,1,2,2,3,3-d6)benzoyl)benzofuran-5-yl)methanesulfonamide Hydrochloride IUPAC Name: N-(2-butyl-3-(4-(3-(butylamino)propoxy-1,1,2,2,3,3-d6)benzoyl)benzofuran-5-yl)methanesulfonamide hydrochloride SMILES: C(=O)(C=1C=2C(OC1CCCC)=CC=C(NS(C)(=O)=O)C2)C3=CC=C(OC(C(C(NCCCC)([2H])[2H])([2H])[2H])([2H])[2H])C=C3.Cl