accessibility menu, dialog, popup

UserName

CAS RN 13478-16-5

IUPAC Name: magnesium(2+) ammonium hexahydrate phosphate
Synonyms: magnesium(2+) ammonium hexahydrate phosphate
Molecular Weight (g/mol): 245.4
Molecular Formula: H16MgNO10P
InChi Key: CKMXBZGNNVIXHC-UHFFFAOYSA-L
SMILES: [NH4+].O.O.O.O.O.O.[Mg++].[O-]P([O-])([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)