accessibility menu, dialog, popup

UserName

CAS RN 13498-08-3

H2OYbOClOOOOClOOOOClOOO

CAS RN 13498-08-3

IUPAC Name: ytterbium(3+) hydrate triperchlorate
Synonyms: ytterbium(3+) hydrate triperchlorate
Molecular Weight (g/mol): 489.41
Molecular Formula: Cl3H2O13Yb
InChi Key: TYYKUTNLJJSRBB-UHFFFAOYSA-K
SMILES: O.[Yb+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results