CAS RN 13535-91-6
CAS RN 13535-91-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(4-Methylquinazolin-2-yl)methanol, TRC
CAS: 13535-91-6 Molecular Formula: C10H10N2O Molecular Weight (g/mol): 174.2 Synonym: 2-Hydroxymethyl-4-methylquinazoline IUPAC Name: (4-methylquinazolin-2-yl)methanol SMILES: Cc1nc(CO)nc2ccccc12