CAS RN 1355244-02-8
CAS RN 1355244-02-8
IUPAC Name:
N-{3-[4-(4-methoxyphenyl)piperazin-1-yl]propyl}-2-{13-methyl-7-oxo-2,8,10-triazatricyclo[7.4.0.0²,⁶]trideca-1(13),3,5,9,11-pentaen-8-yl}acetamide
Synonyms:
N-{3-[4-(4-methoxyphenyl)piperazin-1-yl]propyl}-2-{13-methyl-7-oxo-2,8,10-triazatricyclo[7.4.0.0²,⁶]trideca-1(13),3,5,9,11-pentaen-8-yl}acetamide
Molecular Weight (g/mol):
488.59
Molecular Formula:
C27H32N6O3
InChi Key:
PMTIWRPLQBVEMR-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C=C1)N1CCN(CCCNC(=O)CN2C(=O)C3=CC=CN3C3=C(C)C=CN=C23)CC1
1
–
1
of
1
results
1