accessibility menu, dialog, popup

UserName

CH3H3CCH3OONBHOHO

CAS RN 135884-31-0

IUPAC Name: {1-[(tert-butoxy)carbonyl]-1H-pyrrol-2-yl}boronic acid
Synonyms: 1-(tert-butoxycarbonyl)pyrrol-2-ylboronic acid
Molecular Weight (g/mol): 211.02
Molecular Formula: C9H14BNO4
InChi Key: ZWGMJLNXIVRFRJ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1C=CC=C1B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results