accessibility menu, dialog, popup

UserName

BONFFFK

CAS RN 1359865-98-7

IUPAC Name: potassium trifluoro[4-(piperidine-1-carbonyl)phenyl]boranuide
Synonyms: potassium trifluoro[4-(piperidine-1-carbonyl)phenyl]boranuide
Molecular Weight (g/mol): 295.15
Molecular Formula: C12H14BF3KNO
InChi Key: HAQFTXUNHFCQOC-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)C1=CC=C(C=C1)C(=O)N1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results