accessibility menu, dialog, popup

UserName

CAS RN 136194-77-9

CH3NNNNHO

CAS RN 136194-77-9

IUPAC Name: 3-{23-methyl-14-oxo-3,13,23-triazahexacyclo[14.7.0.0²,¹⁰.0⁴,⁹.0¹¹,¹⁵.0¹⁷,²²]tricosa-1(16),2(10),4,6,8,11(15),17(22),18,20-nonaen-3-yl}propanenitrile
Synonyms: 3-{23-methyl-14-oxo-3,13,23-triazahexacyclo[14.7.0.0²,¹⁰.0⁴,⁹.0¹¹,¹⁵.0¹⁷,²²]tricosa-1(16),2(10),4,6,8,11(15),17(22),18,20-nonaen-3-yl}propanenitrile
Molecular Weight (g/mol): 378.44
Molecular Formula: C24H18N4O
InChi Key: VWVYILCFSYNJHF-UHFFFAOYSA-N
SMILES: CN1C2=C(C=CC=C2)C2=C1C1=C(C3=C2C(=O)NC3)C2=CC=CC=C2N1CCC#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results