accessibility menu, dialog, popup

UserName

OP

CAS RN 13689-19-5

IUPAC Name: (dicyclohexylphosphoroso)cyclohexane
Synonyms: (dicyclohexylphosphoroso)cyclohexane
Molecular Weight (g/mol): 296.44
Molecular Formula: C18H33OP
InChi Key: LEFPWWWXFFNJAA-UHFFFAOYSA-N
SMILES: O=P(C1CCCCC1)(C1CCCCC1)C1CCCCC1
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)