accessibility menu, dialog, popup

UserName

CH3H3CHO

CAS RN 137-32-6

IUPAC Name: 2-methylbutan-1-ol
Synonyms: 2-methyl-1-butanol (1)-2-methylbutan-1-ol active amyl alcohol primary active amyl alcohol active primary amyl alcohol methyl-2-butan-1-ol
Molecular Weight (g/mol): 88.15
Molecular Formula: C5H12O
InChi Key: QPRQEDXDYOZYLA-UHFFFAOYNA-N
SMILES: CCC(C)CO
Boiling Point 
  • (3)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (3)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)