accessibility menu, dialog, popup

UserName

H3CFFF

CAS RN 137019-95-5

IUPAC Name: 3,4,5-trifluoro-4'-(4-pentylcyclohexyl)-1,1'-biphenyl
Synonyms: 3,4,5-trifluoro-4'-(4-pentylcyclohexyl)-1,1'-biphenyl
Molecular Weight (g/mol): 360.46
Molecular Formula: C23H27F3
InChi Key: PRYCYWMMSZSXBK-UHFFFAOYSA-N
SMILES: CCCCCC1CCC(CC1)C1=CC=C(C=C1)C1=CC(F)=C(F)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results