accessibility menu, dialog, popup

UserName

CAS RN 13760-78-6

FFFHo

CAS RN 13760-78-6

IUPAC Name: holmium(3+) trifluoride
Synonyms: holmium(3+) trifluoride
Molecular Weight (g/mol): 221.93
Molecular Formula: F3Ho
InChi Key: FDIFPFNHNADKFC-UHFFFAOYSA-K
SMILES: [F-].[F-].[F-].[Ho+3]
Percent Purity 
  • (6)
  • (2)
Physical Form 
  • (2)
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (6)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)