accessibility menu, dialog, popup

UserName

IIITm

CAS RN 13813-43-9

IUPAC Name: thulium(3+) triiodide
Synonyms: thulium(3+) triiodide
Molecular Weight (g/mol): 549.65
Molecular Formula: I3Tm
InChi Key: LZOMHYVAEHYDST-UHFFFAOYSA-K
SMILES: [I-].[I-].[I-].[Tm+3]
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results