accessibility menu, dialog, popup

UserName

IIILu

CAS RN 13813-45-1

IUPAC Name: lutetium(3+) triiodide
Synonyms: lutetium(3+) triiodide
Molecular Weight (g/mol): 555.68
Molecular Formula: I3Lu
InChi Key: NZOCXFRGADJTKP-UHFFFAOYSA-K
SMILES: [I-].[I-].[I-].[Lu+3]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results