accessibility menu, dialog, popup

UserName

FFFFFFFFFF

CAS RN 138495-42-8

IUPAC Name: 1,1,1,2,2,3,4,5,5,5-decafluoropentane
Synonyms: decafluoropentane
Molecular Weight (g/mol): 252.06
Molecular Formula: C5H2F10
InChi Key: RIQRGMUSBYGDBL-UHFFFAOYNA-N
SMILES: FC(C(F)C(F)(F)C(F)(F)F)C(F)(F)F
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)