CAS RN 13865-20-8
CAS RN 13865-20-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Methyl 2,2-dimethyl-3-oxopropanoate, TRC
CAS: 13865-20-8 Molecular Formula: C6H10O3 Molecular Weight (g/mol): 130.14 IUPAC Name: methyl 2,2-dimethyl-3-oxopropanoate SMILES: COC(=O)C(C)(C)C=O