CAS RN 138804-88-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Methylprednisolone 21-Propionate, TRC
CAS: 138804-88-3 Molecular Formula: C25 H34 O6 Molecular Weight (g/mol): 430.53 Synonym: (6α,11β)-11,17-Dihydroxy-6-methyl-21-(1-oxopropoxy)pregna-1,4-diene-3,20-dione IUPAC Name: [2-[(6S,8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-6,10,13-trimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl]-2-oxo-ethyl] propanoate SMILES: CCC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3C[C@H](C)C4=CC(=O)C=C[C@]4(C)[C@H]3[C@@H](O)C[C@]12C