CAS RN 1391054-57-1
CAS RN 1391054-57-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Alpha-Desmethyl Benoxaprofen Methyl Ester, TRC
CAS: 1391054-57-1 Molecular Formula: C16H12ClNO3 Molecular Weight (g/mol): 301.72 Synonym: 2-(4-Chlorophenyl)-5-benzoxazoleacetic Acid Methyl Ester SMILES: COC(=O)CC1=CC2=C(C=C1)OC(=N2)C3=CC=C(C=C3)Cl