accessibility menu, dialog, popup

UserName

CAS RN 13986-53-3

CoOOOCCCH3CH3H3CH3CCH3H3CH3CCH3H3CCH3CH3H3CHHO

CAS RN 13986-53-3

IUPAC Name: λ²-cobalt(2+) bis(2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate)
Synonyms: λ²-cobalt(2+) bis(2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate)
Molecular Weight (g/mol): 425.48
Molecular Formula: C22H38CoO4
InChi Key: PPQGKVCRZSKSEK-UHFFFAOYSA-L
SMILES: [Co++].CC(C)(C)C([O-])=CC(=O)C(C)(C)C.CC(C)(C)C([O-])=CC(=O)C(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results