accessibility menu, dialog, popup

UserName

KH3COSS

CAS RN 140-89-6

IUPAC Name: potassium (ethoxymethanethioyl)sulfanide
Synonyms: potassium ethyl xanthate
Molecular Weight (g/mol): 160.29
Molecular Formula: C3H5KOS2
InChi Key: JCBJVAJGLKENNC-UHFFFAOYSA-M
SMILES: [K+].CCOC([S-])=S
Molecular Weight (g/mol) 
  • (6)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (6)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)